missing translation for 'onlineSavingsMsg'
Learn More

Bicyclohexyl 99.0+%, TCI America™

Catalog No. B090225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B090225ML 25 mL
B0902100ML 100 mL
B0902500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B090225ML Supplier TCI America Supplier No. B090225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-51-3
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00003815
InChI Key WVIIMZNLDWSIRH-UHFFFAOYSA-N
Synonym bicyclohexyl, 1,1'-bicyclohexyl, dicyclohexyl, bicyclohexane, dicyclohexane, dodecahydrobiphenyl, cyclohexane, cyclohexyl, 1,1'-biphenyl, dodecahydro, 1,1'-bi cyclohexyl, 1, dodecahydro
PubChem CID 7094
IUPAC Name 1,1'-bi(cyclohexane)
SMILES C1CCC(CC1)C1CCCCC1

Specifications

Color Colorless
Boiling Point 239°C
Quantity 25 mL
Formula Weight 166.31
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Bicyclohexyl