Learn More
Bicyclohexyl, 98%
CAS: 92-51-3 | C12H22 | 166.31 g/mol
Supplier: Thermo Scientific Chemicals A1348118
Description
Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Bicyclohexyl | |
| 3°C to 4°C | |
| 226°C to 228°C | |
| C12H22 | |
| MFCD00003815 | |
| 1848266 | |
| Difficult to mix. | |
| C1CCC(CC1)C1CCCCC1 | |
| 166.31 | |
| 166.31 |
| 92-51-3 | |
| 0.864 | |
| 92°C (198°F) | |
| 1.478 | |
| 50 g | |
| bicyclohexyl, 1,1'-bicyclohexyl, dicyclohexyl, bicyclohexane, dicyclohexane, dodecahydrobiphenyl, cyclohexane, cyclohexyl, 1,1'-biphenyl, dodecahydro, 1,1'-bi cyclohexyl, 1, dodecahydro | |
| WVIIMZNLDWSIRH-UHFFFAOYSA-N | |
| cyclohexylcyclohexane | |
| 7094 | |
| 98% |
Chemical Identifiers
| 92-51-3 | |
| 166.31 | |
| WVIIMZNLDWSIRH-UHFFFAOYSA-N | |
| 7094 | |
| C1CCC(CC1)C1CCCCC1 |
| C12H22 | |
| MFCD00003815 | |
| bicyclohexyl, 1,1'-bicyclohexyl, dicyclohexyl, bicyclohexane, dicyclohexane, dodecahydrobiphenyl, cyclohexane, cyclohexyl, 1,1'-biphenyl, dodecahydro, 1,1'-bi cyclohexyl, 1, dodecahydro | |
| cyclohexylcyclohexane |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 202-161-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only