missing translation for 'onlineSavingsMsg'
Learn More

Bicyclohexyl, 98%

Catalog No. AAA1348109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1348109 10 g
AAA1348118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1348109 Supplier Thermo Scientific Chemicals Supplier No. A1348109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 92-51-3 | C12H22 | 166.31 g/mol

Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 92-51-3
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00003815
InChI Key WVIIMZNLDWSIRH-UHFFFAOYSA-N
Synonym bicyclohexyl, 1,1'-bicyclohexyl, dicyclohexyl, bicyclohexane, dicyclohexane, dodecahydrobiphenyl, cyclohexane, cyclohexyl, 1,1'-biphenyl, dodecahydro, 1,1'-bi cyclohexyl, 1, dodecahydro
PubChem CID 7094
IUPAC Name cyclohexylcyclohexane
SMILES C1CCC(CC1)C1CCCCC1

Specifications

Melting Point 3°C to 4°C
Density 0.864
Boiling Point 226°C to 228°C
Flash Point 92°C (198°F)
Refractive Index 1.478
Quantity 10 g
Beilstein 1848266
Solubility Information Difficult to mix.
Formula Weight 166.31
Percent Purity 98%
Chemical Name or Material Bicyclohexyl
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 202-161-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only