missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicyclohexane-4,4'-dione Monoethylene Ketal 98.0+%, TCI America™
$59.65 - $175.22
Chemical Identifiers
| CAS | 56309-94-5 |
|---|---|
| Molecular Formula | C14H22O3 |
| Molecular Weight (g/mol) | 238.327 |
| MDL Number | MFCD00809704 |
| InChI Key | ZNWLFTSPNBLXGL-UHFFFAOYSA-N |
| Synonym | ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl |
| PubChem CID | 810913 |
| IUPAC Name | 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one |
| SMILES | C1CC(=O)CCC1C2CCC3(CC2)OCCO3 |
Chemical Identifiers
| 56309-94-5 | |
| 238.327 | |
| ZNWLFTSPNBLXGL-UHFFFAOYSA-N | |
| 810913 | |
| C1CC(=O)CCC1C2CCC3(CC2)OCCO3 |
| C14H22O3 | |
| MFCD00809704 | |
| ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl | |
| 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one |
Specifications
| 56309-94-5 | |
| White-Yellow | |
| MFCD00809704 | |
| ketoketal, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexanone, 8-4-oxocyclohexyl-1,4-dioxaspiro 4.5 decane, 4-1,4-dioxaspiro 4.5 dec-8-yl cyclohexanone, 4-1,4-dioxaspiro 4.5 decan-8-yl cyclohexan-1-one, bicyclohexane-4,4'-dione monoethylene ketal, 1,1'-bicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'dione monoethylene ketal, dicyclohexane-4,4'-dione monoethylene ketal, cyclohexanone, 4-1,4-dioxaspiro 4.5 dec-8-yl | |
| C1CC(=O)CCC1C2CCC3(CC2)OCCO3 | |
| 238.327 | |
| 238.33 | |
| Crystalline Powder |
| 102°C | |
| C14H22O3 | |
| 5 g | |
| ZNWLFTSPNBLXGL-UHFFFAOYSA-N | |
| 4-(1,4-dioxaspiro[4.5]decan-8-yl)cyclohexan-1-one | |
| 810913 | |
| ≥98.0% (GC) | |
| Bicyclohexane-4,4′-dione Monoethylene Ketal |
Safety and Handling
EINECSNumber : (5)-6425
TSCA : No