missing translation for 'onlineSavingsMsg'
Learn More

[Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers) 97.0+%, TCI America™

Catalog No. p-7122812
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32141G 1 g
B32145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32141G Supplier TCI America Supplier No. B32141G
Only null left

Chemical Identifiers

CAS 18401-43-9
Molecular Formula C13H24O3Si
Molecular Weight (g/mol) 256.417
MDL Number MFCD00167591
InChI Key HHKDWDAAEFGBAC-UHFFFAOYSA-N
Synonym 5-(Triethoxysilyl)-2-norbornene, 5-(Triethoxysilyl)bicyclo[2.2.1]hept-2-ene
PubChem CID 86718
IUPAC Name 5-bicyclo[2.2.1]hept-2-enyl(triethoxy)silane
SMILES CCO[Si](C1CC2CC1C=C2)(OCC)OCC

Specifications

Color Yellow
Boiling Point 104°C
Quantity 1 g
Formula Weight 256.42
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material [Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers)