missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicyclo[1.1.1]pentan-1-amine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 240°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 119.59 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bicyclo[1.1.1]pentan-1-amine Hydrochloride |
Safety and Handling
| TSCA | No |