missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[1.1.1]pentan-1-amine Hydrochloride 98.0+%, TCI America™

Catalog No. b5479200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5479200MG 200 mg
B54791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5479200MG Supplier TCI America Supplier No. B5479200MG

Chemical Identifiers

CAS 22287-35-0
Molecular Formula C5H10ClN
Molecular Weight (g/mol) 119.592
MDL Number MFCD28166320
InChI Key LQKLVOWNBKJRJE-UHFFFAOYSA-N
PubChem CID 54589873
IUPAC Name bicyclo[1.1.1]pentan-3-amine;hydrochloride
SMILES C1C2CC1(C2)N.Cl

Specifications

Melting Point 240°C
Color White
Quantity 200 mg
Formula Weight 119.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bicyclo[1.1.1]pentan-1-amine Hydrochloride