missing translation for 'onlineSavingsMsg'
Learn More

(+)-Bicuculline 98.0+%, TCI America™

Catalog No. B189025MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B189025MG 25 mg
B1890100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B189025MG Supplier TCI America Supplier No. B189025MG

Chemical Identifiers

CAS 485-49-4
Molecular Formula C20H17NO6
Molecular Weight (g/mol) 367.357
MDL Number MFCD00005006
InChI Key IYGYMKDQCDOMRE-ZWKOTPCHSA-N
Synonym +-bicuculline, bicuculline, d-bicuculline, bicculine, bicucullin, bicuculline +, unii-y37615dvkc, 6r-6-5s-6-methyl-5,6,7,8-tetrahydro 1,3 dioxolo 4,5-g isoquinolin-5-yl furo 3,4-e 1,3 benzodioxol-8 6h-one, furo 3,4-e-1,3-benzodioxol-8 6h-one, 6-5,6,7,8-tetrahydro-6-methyl-1,3-dioxolo 4,5-g isoquinolin-5-yl-, r-r*,s*, 6r-6-5s-6-methyl 5,6,7,8-tetrahydro-2h-1,3-dioxolano 4,5-g isoquinolin-5-yl-6-hydro-2h-1,3-dioxoleno 4,5-e isobenzofuran-8-one
PubChem CID 10237
ChEBI CHEBI:3092
IUPAC Name (6R)-6-[(5S)-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-6H-furo[3,4-g][1,3]benzodioxol-8-one
SMILES CN1CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3

Specifications

Melting Point 197°C
Color Yellow
Quantity 25 mg
UN Number 1544
Formula Weight 367.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Bicuculline
RTECSNumber LV0909840
TSCA No