missing translation for 'onlineSavingsMsg'
Learn More

Bicinchoninic Acid Disodium Salt Hydrate 98.0+%, TCI America™

Catalog No. B45091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45091G 1 g
B45095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45091G Supplier TCI America Supplier No. B45091G

Chemical Identifiers

CAS 979-88-4
Molecular Formula C20H10N2Na2O4
Molecular Weight (g/mol) 388.29
MDL Number MFCD00037500
InChI Key AUPXFICLXPLHBB-UHFFFAOYSA-L
Synonym sodium 2,2'-biquinoline-4,4'-dicarboxylate, 2,2'-biquinoline-4,4-dicarboxylic acid disodium salt, 2,2'-biquinoline-4,4'-dicarboxylic acid, disodium salt, sodium bicinchoninate, 2,2'-biquinoline-4,4'-dicarboxylic acid disodium salt, disodium 2,2'-biquinoline-4,4'-dicarboxylate, bicinchoninic acid disodium salt, bca, 2,2'-biquinoline-4,4'-dicarboxylic acid, sodium salt 1:2
PubChem CID 164763
IUPAC Name disodium [2,2'-biquinoline]-4,4'-dicarboxylate
SMILES [Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 388.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bicinchoninic Acid Disodium Salt Hydrate
TSCA Yes
Recommended Storage Refrigerator