missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ Bib Overall 9 oz. Khaki Indura Ultra Soft, Multi
Protective bib overalls provide the highest level of protection in work with electrical exposures and hazards.
Supplier: National Safety Apparel™ C45UTUK254X32
Description
- FR fabric will not melt, drip or ignite
- FR hook and loop fly closure
- FR hook and loop calf closure for easy boot entry
- Inherently flame resistant for FR Protection that will not wash off
- Absorbs moisture
- Quick disconnect and easy to adjust
- Shoulder straps for quick entry
Specifications
| Buckle | |
| Cotton, Nylon | |
| 4X-Large | |
| Open | |
| Standard | |
| CAT 3, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462 |
| Khaki | |
| Stitched | |
| Adjustable | |
| 25 cal/cm² | |
| Flame | |
| Reusable |