missing translation for 'onlineSavingsMsg'
Learn More

Bevantolol Hydrochloride 98.0+%, TCI America™

Catalog No. p-7124060
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5372250MG 250 mg
B53721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5372250MG Supplier TCI America Supplier No. B5372250MG
Only null left

Chemical Identifiers

CAS 42864-78-8
Molecular Formula C20H28ClNO4
Molecular Weight (g/mol) 381.90
MDL Number MFCD00941389
InChI Key FJTKCFSPYUMXJB-UHFFFAOYNA-N
Synonym 1-[[2-(3,4-Dimethoxyphenyl)ethyl]amino]-3-(3-methylphenoxy)-2-propanol Hydrochloride, 1-[(3,4-Dimethoxyphenethyl)amino]-3-(m-tolyloxy)propan-2-ol Hydrochloride
PubChem CID 39324
ChEBI CHEBI:31282
IUPAC Name hydrogen 1-{[2-(3,4-dimethoxyphenyl)ethyl]amino}-3-(3-methylphenoxy)propan-2-ol chloride
SMILES [H+].[Cl-].COC1=CC=C(CCNCC(O)COC2=CC=CC(C)=C2)C=C1OC

Specifications

Melting Point 142°C
Color White
Quantity 250 mg
Formula Weight 381.90
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bevantolol Hydrochloride