missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bevantolol Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 142°C |
| Color | White |
| Quantity | 250 mg |
| Formula Weight | 381.90 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bevantolol Hydrochloride |
Safety and Handling
| TSCA | No |