missing translation for 'onlineSavingsMsg'
Learn More

Betulinol 97.0+%, TCI America™

Catalog No. B0803100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0803100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0803100MG Supplier TCI America Supplier No. B0803100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 473-98-3
Molecular Formula C30H50O2
Molecular Weight (g/mol) 442.728
MDL Number MFCD00016802
InChI Key FVWJYYTZTCVBKE-ROUWMTJPSA-N
Synonym betulin, betulinol, betuline, trochol, betulol, lup-20 29-ene-3b,28-diol, lup-20 29-ene-3beta,28-diol, unii-6w70hn7x7o, lup-20 30-ene-3beta,28-diol, lup-20 29-ene-3,28-diol, 3beta
PubChem CID 72326
ChEBI CHEBI:3086
IUPAC Name (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a-(hydroxymethyl)-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysen-9-ol
SMILES CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)CO

Specifications

Melting Point 253°C
Color White-Yellow
Quantity 100 mg
Formula Weight 442.73
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Betulinol
RTECSNumber OK5755000
TSCA No