missing translation for 'onlineSavingsMsg'
Learn More

Betulinic Acid 97.0+%, TCI America™

Catalog No. B2836100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2836100MG 100 mg
B28361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2836100MG Supplier TCI America Supplier No. B2836100MG

Chemical Identifiers

CAS 472-15-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.711
InChI Key QGJZLNKBHJESQX-FZFNOLFKSA-N
Synonym betulinic acid, mairin, betulic acid, ccris 6748, 3-hydroxylup-20 29-en-28-oic acid, 3beta-hydroxy-20 29-lupaene-28-oic acid, 3beta-hydroxy-lup-20 29-en-28-oic acid, lup-20 29-en-28-oic acid, 3-hydroxy-, 3beta, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid
PubChem CID 64971
ChEBI CHEBI:3087
IUPAC Name (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid
SMILES CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)C(=O)O

Specifications

Melting Point 294°C
Quantity 100 mg
Formula Weight 456.71
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Betulinic Acid