missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Betulin

Chemical Identifiers

CAS 473-98-3
Chemical Name or Material Betulin
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466550010
View Documents
Thermo Scientific™
466550010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466550050
View Documents
Thermo Scientific™
466550050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

473-98-3
Betulin

Specifications

473-98-3
Solubility aq. soln.: .. Other solubilities: soluble in acetic acid
Betulin
1 g
256.0°C to 257.0°C
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-475-5

SDS
Documents

Documents

RUO – Research Use Only