missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bethanechol Chloride 98.0+%, TCI America™
$199.46 - $495.84
Chemical Identifiers
| CAS | 590-63-6 |
|---|---|
| Molecular Formula | C7H17ClN2O2 |
| Molecular Weight (g/mol) | 196.675 |
| MDL Number | MFCD00055224 |
| InChI Key | XXRMYXBSBOVVBH-UHFFFAOYSA-N |
| Synonym | bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone |
| PubChem CID | 11548 |
| ChEBI | CHEBI:3085 |
| IUPAC Name | 2-carbamoyloxypropyl(trimethyl)azanium;chloride |
| SMILES | CC(C[N+](C)(C)C)OC(=O)N.[Cl-] |
Chemical Identifiers
| 590-63-6 | |
| 196.675 | |
| XXRMYXBSBOVVBH-UHFFFAOYSA-N | |
| 11548 | |
| 2-carbamoyloxypropyl(trimethyl)azanium;chloride |
| C7H17ClN2O2 | |
| MFCD00055224 | |
| bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone | |
| CHEBI:3085 | |
| CC(C[N+](C)(C)C)OC(=O)N.[Cl-] |
Specifications
| 216°C | |
| White | |
| MFCD00055224 | |
| XXRMYXBSBOVVBH-UHFFFAOYSA-N | |
| 2-carbamoyloxypropyl(trimethyl)azanium;chloride | |
| 196.675 | |
| CHEBI:3085 | |
| ≥98.0% (T) | |
| Bethanechol Chloride |
| 590-63-6 | |
| C7H17ClN2O2 | |
| bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone | |
| CC(C[N+](C)(C)C)OC(=O)N.[Cl-] | |
| 5 g | |
| 11548 | |
| 196.68 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : BR5425000
TSCA : Yes
Recommended Storage : Refrigerator