missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone 98.0+%, TCI America™

Supplier:  TCI America B18375G

 View more versions of this product

Catalog No. B18375G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Betamethasone
White
MFCD00062969
betamethasone, betadexamethasone, flubenisolone, celestone, betamethazone, rinderon, betacorlan, betacortril, betamamallet, betametasone
CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)CO)O)C)O)F)C
392.467
CHEBI:3077
≥98.0% (HPLC)
378-44-9
C22H29FO5
5 g
UREBDLICKHMUKA-DVTGEIKXSA-N
(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
9782
392.47
Crystalline Powder

Chemical Identifiers

378-44-9
392.467
UREBDLICKHMUKA-DVTGEIKXSA-N
9782
(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
C22H29FO5
MFCD00062969
betamethasone, betadexamethasone, flubenisolone, celestone, betamethazone, rinderon, betacorlan, betacortril, betamamallet, betametasone
CHEBI:3077
CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)CO)O)C)O)F)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TU4000000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions