missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone 98.0+%, TCI America™

Catalog No. B18371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18371G 1 g
B18375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18371G Supplier TCI America Supplier No. B18371G

Chemical Identifiers

CAS 378-44-9
Molecular Formula C22H29FO5
Molecular Weight (g/mol) 392.467
MDL Number MFCD00062969
InChI Key UREBDLICKHMUKA-DVTGEIKXSA-N
Synonym betamethasone, betadexamethasone, flubenisolone, celestone, betamethazone, rinderon, betacorlan, betacortril, betamamallet, betametasone
PubChem CID 9782
ChEBI CHEBI:3077
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)CO)O)C)O)F)C

Specifications

Color White
Quantity 1 g
Formula Weight 392.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Betamethasone
RTECSNumber TU4000000
TSCA No