missing translation for 'onlineSavingsMsg'
Learn More

Betahistine Methanesulfonate 99.0+%, TCI America™

Catalog No. B142425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B142425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B142425G Supplier TCI America Supplier No. B142425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54856-23-4
Molecular Formula C10H20N2O6S2
Molecular Weight (g/mol) 328.398
MDL Number MFCD00143688
InChI Key ZBJJDYGJCNTNTH-UHFFFAOYSA-N
Synonym Betahistine Mesylate, 2-[2-(Methylamino)ethyl]pyridine Methanesulfonate
PubChem CID 198334
IUPAC Name methanesulfonic acid;N-methyl-2-pyridin-2-ylethanamine
SMILES CNCCC1=CC=CC=N1.CS(=O)(=O)O.CS(=O)(=O)O

Specifications

Melting Point 112°C
Color White
Quantity 25 g
Formula Weight 328.40
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Betahistine Methanesulfonate
RTECSNumber UT2970200
TSCA No