missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Sitosterol Acetate (contains Campesterol Acetate) 40.0+%, TCI America™
$142.39 - $368.33
Chemical Identifiers
| CAS | 915-05-9 |
|---|---|
| Molecular Formula | C31H52O2 |
| Molecular Weight (g/mol) | 456.755 |
| MDL Number | MFCD00083497 |
| InChI Key | PBWOIPCULUXTNY-LBKBYZTLSA-N |
| PubChem CID | 5354503 |
| ChEBI | CHEBI:69433 |
| IUPAC Name | [(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate |
| SMILES | CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C)C(C)C |
Chemical Identifiers
| 915-05-9 | |
| 456.755 | |
| PBWOIPCULUXTNY-LBKBYZTLSA-N | |
| CHEBI:69433 | |
| CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C)C(C)C |
| C31H52O2 | |
| MFCD00083497 | |
| 5354503 | |
| [(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate |
Specifications
| 915-05-9 | |
| White | |
| MFCD00083497 | |
| PBWOIPCULUXTNY-LBKBYZTLSA-N | |
| [(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate | |
| 5354503 | |
| 456.76 | |
| Crystalline Lumps |
| 133°C | |
| C31H52O2 | |
| 1 g | |
| CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C)C(C)C | |
| 456.755 | |
| CHEBI:69433 | |
| ≥40.0% (GC) | |
| beta-Sitosterol Acetate (contains Campesterol Acetate) |
Safety and Handling
TSCA : No