missing translation for 'onlineSavingsMsg'
Learn More

beta-Sitosterol Acetate (contains Campesterol Acetate) 40.0+%, TCI America™

Catalog No. S02211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02211G 1 g
S02215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02211G Supplier TCI America Supplier No. S02211G

Chemical Identifiers

CAS 915-05-9
Molecular Formula C31H52O2
Molecular Weight (g/mol) 456.755
MDL Number MFCD00083497
InChI Key PBWOIPCULUXTNY-LBKBYZTLSA-N
PubChem CID 5354503
ChEBI CHEBI:69433
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMILES CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C)C(C)C

Specifications

Melting Point 133°C
Color White
Quantity 1 g
Formula Weight 456.76
Percent Purity ≥40.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material beta-Sitosterol Acetate (contains Campesterol Acetate)