missing translation for 'onlineSavingsMsg'
Learn More

(-)-beta-Pinene 94.0+%, TCI America™

Supplier:  TCI America P0441500ML

 View more versions of this product

Catalog No. P0441500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(-)-beta-Pinene
Yellow
C10H16
500 mL
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
CC1(C2CCC(=C)C1C2)C
136.238
CHEBI:28359
≥94.0% (GC)
18172-67-3
165°C
MFCD00063635
3295
WTARULDDTDQWMU-IUCAKERBSA-N
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
440967
136.24
Liquid

Chemical Identifiers

18172-67-3
136.238
WTARULDDTDQWMU-IUCAKERBSA-N
440967
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
C10H16
MFCD00063635
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
CHEBI:28359
CC1(C2CCC(=C)C1C2)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0593&(8)-0497

RTECSNumber : DT5077000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions