missing translation for 'onlineSavingsMsg'
Learn More

(-)-beta-Pinene 94.0+%, TCI America™

Catalog No. P044125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P044125ML 25 mL
P0441500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P044125ML Supplier TCI America Supplier No. P044125ML

Chemical Identifiers

CAS 18172-67-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00063635
InChI Key WTARULDDTDQWMU-IUCAKERBSA-N
Synonym --beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
PubChem CID 440967
ChEBI CHEBI:28359
IUPAC Name (1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
SMILES CC1(C2CCC(=C)C1C2)C

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 mL
UN Number 3295
Formula Weight 136.24
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-beta-Pinene
EINECSNumber (4)-0593&(8)-0497
RTECSNumber DT5077000
TSCA Yes