missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide Adenine Dinucleotide Phosphate Sodium Salt Hydrate, oxidized form 90.0+%, TCI America™

Catalog No. C0379100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0379100MG 100 mg
C03791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0379100MG Supplier TCI America Supplier No. C0379100MG

[for Biochemical Research]

Chemical Identifiers

CAS 1184-16-3
Molecular Formula C21H27N7NaO17P3
Molecular Weight (g/mol) 765.39
MDL Number MFCD00036973
InChI Key JNUMDLCHLVUHFS-QYZPTAICSA-M
Synonym nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
PubChem CID 131845262
IUPAC Name sodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)oxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
SMILES [Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Melting Point 178°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 100 mg
Formula Weight 766.40
Percent Purity ≥90.0% (HPLC)
Chemical Name or Material beta-Nicotinamide Adenine Dinucleotide Phosphate Sodium Salt Hydrate, oxidized form [for Biochemical
TSCA No
Recommended Storage Freezer