missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate, 97%

Catalog No. AAJ60387ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60387ME 500 mg
AAJ60387MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60387ME Supplier Thermo Scientific Chemicals Supplier No. J60387ME

C21H26N7Na4O17P3·xH2O

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

Solubility
It is soluble in 10 mM NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C Light sensitive. Hygroscopic

Chemical Identifiers

Molecular Formula C21H26N7Na4O17P3·xH2O
MDL Number MFCD07366294
Synonym beta-NADPH tetrasodium salt tetrahydrate

Specifications

Color White
Quantity 500 mg
Solubility Information It is soluble in 10 mM NaOH.
Formula Weight 905.42 (833.35 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only