Learn More
beta-Nicotinamide adenine dinucleotide phosphate disodium salt, 97%
Description
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.
Solubility
Soluble in water (50 mg/ml), 0.001% aqueous buffer, pH 7.0, methanol, and ethanol (less soluble). Insoluble in ether, and ethyl acetate.
Notes
Protect from light and moisture. Keep cool and dry. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Store under inert gas.
Specifications
Specifications
| Color | White |
| Quantity | 250 mg |
| Solubility Information | Soluble in water (50mg/ml),0.001% aqueous buffer,pH 7.0,methanol,and ethanol (less soluble). Insoluble in ether,and ethyl acetate. |
| Formula Weight | 787.37 |
| Percent Purity | 97% |
| Physical Form | Powder |
| Chemical Name or Material | beta-Nicotinamide adenine dinucleotide phosphate disodium salt |
Safety and Handling
| EINECSNumber | 246-129-8 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only