missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide Adenine Dinucleotide Hydrate, oxidized form 95.0+%, TCI America™

Catalog No. D09191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09191G 1 g
D09195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09191G Supplier TCI America Supplier No. D09191G

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name 1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 663.43
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material beta-Nicotinamide Adenine Dinucleotide Hydrate, oxidized form [for Biochemical Research]
RTECSNumber UU3450000
TSCA Yes
Recommended Storage Refrigerator