missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Nicotinamide Adenine Dinucleotide Disodium Salt Hydrate, reduced form [for Biochemical Research], TCI America™
$57.79 - $272.16
Chemical Identifiers
| CAS | 606-68-8 |
|---|---|
| Molecular Formula | C21H27N7Na2O14P2 |
| Molecular Weight (g/mol) | 709.41 |
| MDL Number | MFCD00036200 |
| InChI Key | QRGNQKGQENGQSE-WUEGHLCSSA-L |
| Synonym | beta-nadh disodium salt |
| PubChem CID | 131673989 |
| IUPAC Name | disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
| SMILES | [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
Chemical Identifiers
| 606-68-8 | |
| 709.41 | |
| QRGNQKGQENGQSE-WUEGHLCSSA-L | |
| 131673989 | |
| [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
| C21H27N7Na2O14P2 | |
| MFCD00036200 | |
| beta-nadh disodium salt | |
| disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
Specifications
| 606-68-8 | |
| C21H27N7Na2O14P2 | |
| 100 mg | |
| QRGNQKGQENGQSE-WUEGHLCSSA-L | |
| disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate | |
| 131673989 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00036200 | |
| beta-nadh disodium salt | |
| [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
| 709.41 | |
| 709.41 | |
| beta-Nicotinamide Adenine Dinucleotide Disodium Salt Hydrate, reduced form [for Biochemical Research |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer