missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
beta-Nicotinamide adenine dinucleotide, disodium salt, hydrate, 95+%, reduced form, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC271100250
Specifications
0.79 to 0.85 (A250/A260), 0.21 to 0.25 (A280/A260) | |
Authentic | |
8% Max. (K.F.) | |
140.0°C to 142.0°C | |
Reduced Form | |
95.0 | |
95% min. (HPLC) | |
MFCD00036200 | |
Solubility in water: soluble | |
[HH].[HH].C1C=CN(C=C1C(=O)N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O.[Na].[Na] | |
709.4 | |
709.4 | |
95+% |
β-Nicotinamide adenine dinucleotide, disodium salt, hydrate | |
Glass bottle | |
Yellow | |
25g | |
1949720-50-6 | |
100.0 | |
C21H27N7Na2O14P2·H2O | |
beta-nadh disodium salt | |
PSTAPGCXUOIFPQ-ITGWJZMWSA-N | |
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium | |
131673989 | |
Powder |
Chemical Identifiers
1949720-50-6 | |
709.4 | |
PSTAPGCXUOIFPQ-ITGWJZMWSA-N | |
131673989 | |
[HH].[HH].C1C=CN(C=C1C(=O)N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O.[Na].[Na] |
C21H27N7Na2O14P2·H2O | |
MFCD00036200 | |
beta-nadh disodium salt | |
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |