missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide, 97%

Catalog No. p-7042385
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6233703 1 g
AAJ6233706 5 g
AAJ6233714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6233703 Supplier Thermo Scientific Chemicals Supplier No. J6233703

CAS: 53-84-9 | C21H27N7O14P2 | 663.43 g/mol

NAD+, Free Acid is a major electron acceptor molecule in biological oxidation reactions. It is also used as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NAD+, Free Acid is a major electron acceptor molecule in biological oxidation reactions. It is also used as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling,.

Solubility
Soluble in water at 50mg/ml

Notes
b-NAD is very hygroscopic and should be stored desiccated. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C.

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 1 g
Beilstein 3584133
Merck Index 14,6344
Solubility Information Soluble in water at 50mg/ml
Formula Weight 663.43
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material beta-Nicotinamide adenine dinucleotide
EINECSNumber 200-184-4
RTECSNumber UU3450000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only