missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Methallyl Alcohol 98.0+%, TCI America™
Supplier: TCI America M0091500ML
Specifications
| beta-Methallyl Alcohol | |
| Colorless | |
| C4H8O | |
| 500 mL | |
| methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol | |
| CC(=C)CO | |
| 72.107 | |
| 72.11 | |
| Liquid |
| 513-42-8 | |
| 113°C | |
| MFCD00004737 | |
| 2614 | |
| BYDRTKVGBRTTIT-UHFFFAOYSA-N | |
| 2-methylprop-2-en-1-ol | |
| 10557 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 513-42-8 | |
| 72.107 | |
| BYDRTKVGBRTTIT-UHFFFAOYSA-N | |
| 10557 | |
| CC(=C)CO |
| C4H8O | |
| MFCD00004737 | |
| methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol | |
| 2-methylprop-2-en-1-ol |
Safety and Handling
EINECSNumber : (2)-3872
RTECSNumber : UD5250000
TSCA : Yes