missing translation for 'onlineSavingsMsg'
Learn More

beta-Methallyl Alcohol 98.0+%, TCI America™

Supplier:  TCI America M009125ML

 View more versions of this product

Catalog No. M009125ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

beta-Methallyl Alcohol
Colorless
C4H8O
25 mL
methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
CC(=C)CO
72.107
72.11
Liquid
513-42-8
113°C
MFCD00004737
2614
BYDRTKVGBRTTIT-UHFFFAOYSA-N
2-methylprop-2-en-1-ol
10557
≥98.0% (GC)

Chemical Identifiers

513-42-8
72.107
BYDRTKVGBRTTIT-UHFFFAOYSA-N
10557
CC(=C)CO
C4H8O
MFCD00004737
methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
2-methylprop-2-en-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3872

RTECSNumber : UD5250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions