missing translation for 'onlineSavingsMsg'
Learn More

beta-Methallyl Alcohol 98.0+%, TCI America™

Catalog No. M00915ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M00915ML 5 mL
M009125ML 25 mL
M0091500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M00915ML Supplier TCI America Supplier No. M00915ML
Only null left

Chemical Identifiers

CAS 513-42-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00004737
InChI Key BYDRTKVGBRTTIT-UHFFFAOYSA-N
Synonym methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
PubChem CID 10557
IUPAC Name 2-methylprop-2-en-1-ol
SMILES CC(=C)CO

Specifications

Color Colorless
Boiling Point 113°C
Quantity 5 mL
UN Number 2614
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Methallyl Alcohol
EINECSNumber (2)-3872
RTECSNumber UD5250000
TSCA Yes