missing translation for 'onlineSavingsMsg'
Learn More

beta-L-Ribofuranose 1-Acetate 2,3,5-Tribenzoate 98.0+%, TCI America™

Catalog No. R00801G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00801G Supplier TCI America Supplier No. R00801G
Only null left

Chemical Identifiers

CAS 3080-30-6
Molecular Formula C28H24O9
Molecular Weight (g/mol) 504.491
MDL Number MFCD04114304
InChI Key GCZABPLTDYVJMP-TVQWTUMOSA-N
Synonym 1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-L-ribofuranose
PubChem CID 10973190
IUPAC Name [(2S,3S,4S,5R)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
SMILES CC(=O)OC1C(C(C(O1)COC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 504.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material beta-L-Ribofuranose 1-Acetate 2,3,5-Tribenzoate