missing translation for 'onlineSavingsMsg'
Learn More

beta-Ionone 95.0+%, TCI America™

Catalog No. p-7131199
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I007725ML 25 mL
I0077100ML 100 mL
I0077500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I007725ML Supplier TCI America Supplier No. I007725ML
Only null left

Chemical Identifiers

CAS 14901-07-6
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.30
MDL Number MFCD00001549
InChI Key PSQYTAPXSHCGMF-BQYQJAHWSA-N
Synonym beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
PubChem CID 638014
ChEBI CHEBI:32325
IUPAC Name (3E)-4-(2,6,6-trimethylcyclohex-1-en-1-yl)but-3-en-2-one
SMILES CC(=O)\C=C\C1=C(C)CCCC1(C)C

Specifications

Color Yellow
Boiling Point 151°C
Quantity 25 mL
Formula Weight 192.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Ionone
EINECSNumber (3)-2387
RTECSNumber EN0350000
TSCA Yes