missing translation for 'onlineSavingsMsg'
Learn More

beta-Ionone, 96%

Catalog No. p-7021172
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Catalog No. Quantity
AAA1340022 100 g
AAA1340036 500 g
2 options

Catalog No. AAA1340022

Supplier: Thermo Scientific Chemicals A1340022

Only null left

CAS: 79-77-6 | C13H20O | 192.302 g/mol

It is an important fragrance chemical used in perfumery. The combination of α-ionone and β-ionone is characteristic of the scent of violets and used with other components in flavoring to recreate their scent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important fragrance chemical used in perfumery. The combination of α-ionone and β-ionone is characteristic of the scent of violets and used with other components in flavoring to recreate their scent.

Solubility
Soluble in water (0.11 mg/mI at 20°C).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 79-77-6
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00001549
InChI Key PSQYTAPXSHCGMF-BQYQJAHWSA-N
Synonym beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
PubChem CID 638014
ChEBI CHEBI:32325
IUPAC Name (E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one
SMILES CC1=C(C(CCC1)(C)C)C=CC(=O)C

Specifications

Melting Point -49°C
Density 0.943
Boiling Point 126°C to 128°C (12 mmHg)
Flash Point 112°C (233°F)
Refractive Index 1.52
Quantity 100 g
Beilstein 1909544
Merck Index 14,5056
Solubility Information Soluble in water (0.11mg/mI at 20°C).
Formula Weight 192.3
Percent Purity 96%
Chemical Name or Material beta-Ionone
Show More Show Less
EINECSNumber 201-224-3
RTECSNumber EN0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only