missing translation for 'onlineSavingsMsg'
Learn More

beta-Glycerophosphate, 200mM soln.

Catalog No. p-7042320
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62121AD 50 mL
AAJ62121AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62121AD Supplier Thermo Scientific Chemicals Supplier No. J62121AD
Only null left

CAS: 819-83-0 | C3H9Na2O6P | 218.052 g/mol

β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 819-83-0
Molecular Formula C3H7Na2O6P
Molecular Weight (g/mol) 216.04
MDL Number MFCD00002135 MFCD28134611 MFCD00149083 MFCD00150339 MFCD00002135
InChI Key AVPCPPOOQICIRJ-UHFFFAOYSA-L
Synonym glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2o
PubChem CID 131860121
IUPAC Name disodium 2-(phosphonatooxy)propane-1,3-diol
SMILES [Na+].[Na+].OCC(CO)OP([O-])([O-])=O

Specifications

Color Colorless
Quantity 50 mL
Solubility Information Soluble in water.
Formula Weight 216.04
Concentration or Composition (by Analyte or Components) 200mM soln.
Physical Form Liquid
Chemical Name or Material beta-Glycerophosphate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only