Learn More
beta-D-Glucose pentaacetate, 98%
CAS: 604-69-3 | C16H22O11 | 390.341 g/mol
Supplier: Thermo Scientific Chemicals A125260B
Description
β-D-Glucose Pentaacetate is used in biochemical reaction and also used as an active pharmaceutical intermediate. Further, it is used to stimulate insulin release in rat pancreatic islets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsβ-D-Glucose Pentaacetate is used in biochemical reaction and also used as an active pharmaceutical intermediate. Further, it is used to stimulate insulin release in rat pancreatic islets.
Solubility
Soluble in chloroform and methanol. Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
| beta-D-Glucose pentaacetate | |
| 130°C to 132°C | |
| MFCD00006597 | |
| 98851 | |
| Soluble in chloroform and methanol. Insoluble in water. | |
| +5° (c=5 in Chloroform) | |
| [(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate | |
| 2724702 | |
| 98% |
| 604-69-3 | |
| C16H22O11 | |
| 1000 g | |
| beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β | |
| LPTITAGPBXDDGR-IBEHDNSVSA-N | |
| CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C | |
| 390.341 | |
| 390.34 |
Chemical Identifiers
| 604-69-3 | |
| 390.341 | |
| LPTITAGPBXDDGR-IBEHDNSVSA-N | |
| 2724702 | |
| CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C |
| C16H22O11 | |
| MFCD00006597 | |
| beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β | |
| [(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate |
Safety and Handling
EINECSNumber : 210-074-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only