missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Glucosamine pentaacetate, 96%

Catalog No. AAL0902003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0902003 1 g
AAL0902006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0902003 Supplier Thermo Scientific Chemicals Supplier No. L0902003

CAS: 7772-79-4 | C16H23NO10 | 389.357 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7772-79-4
Molecular Formula C16H23NO10
Molecular Weight (g/mol) 389.357
MDL Number MFCD00006595
InChI Key OVPIZHVSWNOZMN-OXGONZEZSA-N
Synonym n-acetyl-beta-d-glucosamine tetraacetate, 2-acetamido-1,3,4,6-tetra-o-acetyl-2-deoxy-beta-d-glucopyranose, beta-d-glucosamine pentaacetate, 2-acetamido-2-deoxy-beta-d-glucopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-5-acetamido-3,4,6-triacetoxy-tetrahydropyran-2-yl methyl acetate, beta-d-galactosamine pentaacetate, 2-acetylamino-2-deoxy-beta-d-galactopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-3,4,6-tris acetyloxy-5-acetamidooxan-2-yl methyl acetate, n-acetyl-, a-d-glucosamine tetraacetate, glc-n-ac free
PubChem CID 99461
IUPAC Name [(2R,3S,4R,5R,6S)-5-acetamido-3,4,6-triacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)NC1C(C(C(OC1OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 184°C to 189°C
Quantity 1 g
Beilstein 98835
Optical Rotation +4° (c=10 in Chloroform)
Formula Weight 389.37
Percent Purity 96%
Chemical Name or Material beta-D-Glucosamine pentaacetate
EINECSNumber 231-865-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only