missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Gentiobiose, 98%

Catalog No. p-7043272
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66750MC 100 mg
AAJ66750ME 500 mg
AAJ6675003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66750MC Supplier Thermo Scientific Chemicals Supplier No. J66750MC

CAS: 554-91-6 | C12H22O11

Gentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 554-91-6
Molecular Formula C12H22O11
MDL Number MFCD00198056
Synonym 6-O-(beta-D-Glucopyranosyl)-D-glucopyranose; Amygdalose

Specifications

Color White
Quantity 100 mg
Beilstein 93354
Solubility Information Slightly soluble in water.
Formula Weight 342.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material beta-D-Gentiobiose
EINECSNumber 209-074-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only