missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Galactose pentaacetate, 98%

Catalog No. p-7081955
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1216314 25 g
AAA1216322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1216314 Supplier Thermo Scientific Chemicals Supplier No. A1216314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4163-60-4 | C16H22O11 | 390.341 g/mol

It is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediates.

Solubility
Soluble in methanol (50 mg/ml), chloroform, and water.

Notes
Store at room temperature. Store away from oxidizing agent.

Chemical Identifiers

CAS 4163-60-4
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD00063259
InChI Key LPTITAGPBXDDGR-LYYZXLFJSA-N
Synonym beta-d-galactose pentaacetate, penta-o-acetyl-beta-d-galactopyranose, b-d-galactose pentaacetate, beta-d-galactopyranose pentaacetate, beta-d-galactosepentaacetate, beta-d-galactopyranose, pentaacetate, 2s,3r,4s,5s,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 1,2,3,4,6-penta-o-acetyl-beta-d-galactose, 1,2,3,4,6-penta-o-acetyl-beta-d-galactopyranose, 2r,3s,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate
PubChem CID 94752
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 142°C to 146°C
Quantity 25 g
Beilstein 98853
Solubility Information Soluble in methanol (50mg/ml),chloroform,and water.
Optical Rotation +24° (c=4 in chloroform)
Formula Weight 390.34
Percent Purity 98%
Chemical Name or Material beta-D-Galactose pentaacetate
EINECSNumber 224-008-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only