missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclodextrin

Catalog No. AAJ6316114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6316114 25 g
AAJ6316122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6316114 Supplier Thermo Scientific Chemicals Supplier No. J6316114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7585-39-9 | C42H70O35 | 1134.99 g/mol

Use to solubilize non-polar compounds such as fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomersβ-Cyclodextrin is used with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins. It is an active ingredient of household odor eliminator. It is also used in personal care products like toothpastes, skin creams and dusting powders. It finds applications in the cosmetic industry for products like detergents and perfumes for the controlled release of fragrances. Further, it is used to produce HPLC columns allowing chiral enantiomers separation. In addition to this, it is used to decrease the level of cholesterol in milk fat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use to solubilize non-polar compounds such as fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomersβ-Cyclodextrin is used with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins. It is an active ingredient of household odor eliminator. It is also used in personal care products like toothpastes, skin creams and dusting powders. It finds applications in the cosmetic industry for products like detergents and perfumes for the controlled release of fragrances. Further, it is used to produce HPLC columns allowing chiral enantiomers separation. In addition to this, it is used to decrease the level of cholesterol in milk fat.

Solubility
Soluble in water and ammonium hydroxide.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7585-39-9
Molecular Formula C42H70O35
Molecular Weight (g/mol) 1134.99
MDL Number MFCD00078139
InChI Key WHGYBXFWUBPSRW-UHFFFAOYNA-N
Synonym cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose
PubChem CID 131707246
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Melting Point 290°C (decomposition)
Color White
Quantity 25 g
Beilstein 78623
Merck Index 14,2718
Solubility Information Soluble in water and ammonium hydroxide.
Optical Rotation +162° (c=1.5 in Water)
Formula Weight 1134.98
Physical Form Powder
Chemical Name or Material beta-Cyclodextrin
EINECSNumber 231-493-2
RTECSNumber GU2293000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only