missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Cyclodextrin 98.0+%, TCI America™
Supplier: TCI America C0900100G
| Quantity | 100 g |
|---|
Chemical Identifiers
| 7585-39-9 | |
| 1134.99 | |
| WHGYBXFWUBPSRW-UHFFFAOYNA-N | |
| 131707246 | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
| C42H70O35 | |
| MFCD00078139 | |
| cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose | |
| 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol |
Specifications
| beta-Cyclodextrin | |
| White-Yellow | |
| 100 g | |
| cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
| 1134.99 | |
| 1134.99 | |
| Crystalline Powder |
| 7585-39-9 | |
| C42H70O35 | |
| MFCD00078139 | |
| WHGYBXFWUBPSRW-UHFFFAOYNA-N | |
| 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol | |
| 131707246 | |
| ≥98.0% (HPLC) |
Safety and Handling
EINECSNumber : (8)-0101
RTECSNumber : GU2293000
TSCA : Yes