missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclocitral, 90+%

Catalog No. AAB2570009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2570009 10 g
AAB2570018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2570009 Supplier Thermo Scientific Chemicals Supplier No. B2570009

CAS: 432-25-7 | C10H16O | 152.24 g/mol

beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents.

Chemical Identifiers

CAS 432-25-7
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00079078
InChI Key MOQGCGNUWBPGTQ-UHFFFAOYSA-N
Synonym beta-cyclocitral, b-cyclocitral, 1-cyclohexene-1-carboxaldehyde, 2,6,6-trimethyl, .beta.-cyclocitral, 2,6,6-trimethyl-1-cyclohexene-1-carboxaldehyde, unii-77y0u2x29g, cyclocitral, alpha beta-cyclocitral, 2,6,6-trimethylcyclohex-1-ene-1-carbaldehyde, 1-formyl-2,6,6-trimethyl-1-cyclohexene
PubChem CID 9895
ChEBI CHEBI:53177
IUPAC Name 2,6,6-trimethylcyclohexene-1-carbaldehyde
SMILES CC1=C(C=O)C(C)(C)CCC1

Specifications

Density 0.944
Boiling Point 62°C to 63°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.498
Quantity 10 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,dichloromethane,ethyl acetate and methanol. Insoluble in water.
Formula Weight 152.23
Percent Purity ≥90%
Chemical Name or Material beta-Cyclocitral
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-081-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only