missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Beta Cyanoethyl Fastphoramidite™, dG(dmf)

Catalog No. p-7083021
Encompass
Change view
Click to view available options
Quantity:
1 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401183 1 g
401165 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 401183 Supplier Applied Biosystems™ Supplier No. 401183
Only null left

The dG(dmf) Beta Cyanoethyl Fastphoramidite™ is used as a component of a fast deprotecting chemistry set including dA(Bz), dC(Bz) and T. FastPhoramidite™ series deprotects in 1 hour at 65°C.

High QualityOur phosphoramidites benefit from proprietary manufacturing processes that yield high-purity products. In synthesis they perform at a correspondingly high level, producing pure, High Quality DNA at all scales. We guarantee that 5'-dimethoxytrityl nucleoside β-cyanoethyl phosphoramidites, monomer intermediates for licensed β-cyanoethyl phosphoramidite chemistry, will produce DNA that is at least 99.8% chemically authentic when used with recommended synthesis cycles.Fast ResultsFastPhoramidite™ reagents offer fast deprotection for primer-length oligonucleotides. Simply substituting Fast G, dG(dmf), for our standard dG(iBu) shortens deprotection at 55°C from eight hours to two. Elevating the temperature to 65°C further cuts deprotection to 1 hour.Dry phosphoroamidites (powder form) are guaranteed for one year from the date of shipment. Lifetime of diluted phosphoramidites in residence on the instrument is two weeks when used with CPG synthesis supports, and three weeks when used with polystyrene supports.
  • Proprietary manufacturing processes yield high-purity β-cyanoethyl phosphoramidites, which in turn guarantee production of high-quality DNA at all scales.
  • FastPhoramidite™ reagents deprotect in one hour at 65°C to shorten overall oligonucleotide production times and costs.
  • β-cyanoethyl phosphoramidites are use-tested and optimized for use on the ABI synthesizers to guarantee you high-quality nucleic acid synthesis.

Specifications

Format Powder
For Use With (Equipment) 3900 DNA Synthesizer
Product Type dG(dmf) beta CEP
Content And Storage Store in freezer (-5 to -30°C).
Nucleoside Type Deoxyguanosine (dG)
Reagent Features Fast deprotection
Shipping Condition Room Temperature
Quantity 1 g
Reagent Type Phosphoramidite
Nucleic Acid Type DNA

For Research Use Only. Not for use in diagnostic procedures.