missing translation for 'onlineSavingsMsg'
Learn More

(-)-beta-Citronellol 95.0+%, TCI America™

Supplier:  TCI America C146625ML

Catalog No. C146625ML


Only null left

Description

Description

Specifications

Specifications

(-)-beta-Citronellol
Colorless
C10H20O
25 mL
QMVPMAAFGQKVCJ-JTQLQIEISA-N
(3S)-3,7-dimethyloct-6-en-1-ol
7793
156.27
Liquid
7540-51-4
118°C
MFCD00063214
--citronellol, s-3,7-dimethyloct-6-en-1-ol, --beta-citronellol, s---citronellol, beta-rhodinol, s---beta-citronellol, s-citronellol, --3,7-dimethyloct-6-en-1-ol, 3s-3,7-dimethyloct-6-en-1-ol, unii-8rsy5y5658
CC(CCC=C(C)C)CCO
156.269
CHEBI:88
≥95.0% (GC)

Chemical Identifiers

7540-51-4
156.269
QMVPMAAFGQKVCJ-JTQLQIEISA-N
7793
(3S)-3,7-dimethyloct-6-en-1-ol
C10H20O
MFCD00063214
--citronellol, s-3,7-dimethyloct-6-en-1-ol, --beta-citronellol, s---citronellol, beta-rhodinol, s---beta-citronellol, s-citronellol, --3,7-dimethyloct-6-en-1-ol, 3s-3,7-dimethyloct-6-en-1-ol, unii-8rsy5y5658
CHEBI:88
CC(CCC=C(C)C)CCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0258

RTECSNumber : RH3400000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions