missing translation for 'onlineSavingsMsg'
Learn More

(-)-beta-Citronellol, 97%

Catalog No. p-7040407
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6017403 1 g
AAH6017406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6017403 Supplier Thermo Scientific Chemicals Supplier No. H6017403
Only null left

CAS: 7540-51-4 | C10H20O | 156.269 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7540-51-4
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00063214
InChI Key QMVPMAAFGQKVCJ-JTQLQIEISA-N
Synonym --citronellol, s-3,7-dimethyloct-6-en-1-ol, --beta-citronellol, s---citronellol, beta-rhodinol, s---beta-citronellol, s-citronellol, --3,7-dimethyloct-6-en-1-ol, 3s-3,7-dimethyloct-6-en-1-ol, unii-8rsy5y5658
PubChem CID 7793
ChEBI CHEBI:88
IUPAC Name (3S)-3,7-dimethyloct-6-en-1-ol
SMILES CC(CCC=C(C)C)CCO

Specifications

Density 0.856
Boiling Point 225°C to 226°C
Flash Point 99°C (210°F)
Refractive Index 1.456
Quantity 1 g
Beilstein 1721505
Optical Rotation −5.3° (Neat)
Formula Weight 156.27
Percent Purity 97%
Chemical Name or Material (-)-beta-Citronellol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-415-7
RTECSNumber RH3404000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only