missing translation for 'onlineSavingsMsg'
Learn More

beta-Caryophyllene 90.0+%, TCI America™

Catalog No. C079625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C079625ML 25 mL
C0796100ML 100 mL
C0796500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C079625ML Supplier TCI America Supplier No. C079625ML

Chemical Identifiers

CAS 87-44-5
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00075925
InChI Key NPNUFJAVOOONJE-IZZDOVSWSA-N
Synonym --caryophyllene, caryophyllene, l-caryophyllene, trans-caryophyllene, e-.beta.-caryophyllene, .beta.-caryophyllene,-, caryophyllene ,alpha + beta mixt., + e-caryophyllene, e-.beta.-caryophylene, 8-methylene-4,11-trimethyl bicyclo 7.2.0 undec-4-ene
PubChem CID 5354499
IUPAC Name (4E)-4,11,11-trimethyl-8-methylidenebicyclo[7.2.0]undec-4-ene
SMILES CC1=CCCC(=C)C2CC(C2CC1)(C)C

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 mL
Formula Weight 204.36
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Caryophyllene
EINECSNumber (4)-0592
RTECSNumber DT8400000
TSCA Yes