missing translation for 'onlineSavingsMsg'
Learn More

beta-Carotene 97.0+%, TCI America™

Catalog No. C05605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05605G 5 g
C05601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05605G Supplier TCI America Supplier No. C05605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

beta-Carotene 97.0+%

Chemical Identifiers

CAS 7235-40-7
Molecular Formula C40H56
Molecular Weight (g/mol) 536.89
MDL Number MFCD00001556
InChI Key OENHQHLEOONYIE-JLTXGRSLSA-N
Synonym beta-carotene, beta carotene, betacarotene, beta,beta-carotene, provitamin a, solatene, carotaben, provatene, all-trans-beta-carotene, serlabo
PubChem CID 5280489
ChEBI CHEBI:17579
IUPAC Name 1,3,3-trimethyl-2-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyl-18-(2,6,6-trimethylcyclohex-1-en-1-yl)octadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl]cyclohex-1-ene
SMILES C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C

Specifications

Melting Point 184°C
Quantity 5 g
Formula Weight 536.89
Percent Purity ≥97.0% (E)
Physical Form Crystalline Powder
Chemical Name or Material beta-Carotene
EINECSNumber (8)-0496&(9)-0302
RTECSNumber FI0329500
TSCA Yes