missing translation for 'onlineSavingsMsg'
Learn More

beta-Butyrolactone, TCI America™

Catalog No. p-7118733
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B090125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B090125ML Supplier TCI America Supplier No. B090125ML
Only null left

Chemical Identifiers

CAS 3068-88-0
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00005170
InChI Key GSCLMSFRWBPUSK-UHFFFAOYSA-N
Synonym 4-Methyl-2-oxetanone, beta-Methylpropiolactone
PubChem CID 18303
ChEBI CHEBI:82296
IUPAC Name 4-methyloxetan-2-one
SMILES CC1CC(=O)O1

Specifications

Melting Point -44°C
Color Yellow
Boiling Point 87°C
Quantity 25 mL
UN Number 1993
Formula Weight 86.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Butyrolactone
EINECSNumber (5)-0012
RTECSNumber RQ8050000
TSCA Yes